1-Bromo-2-chloronaphthalene 98%

Supplier: Ambeed
Warning

A113638-1G A113638-250MG A113638-5G
77108-140EA 44.29 USD
77108-140 77108-142 77108-144
1-Bromo-2-chloronaphthalene 98%
1-Bromo-2-chloronaphthalene

Precaución: for Research Use Only

Formula: C₁₀H₆BrCl
MDL Number: MFCD17012389
CAS Number: 71436-66-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR