6-Chloro-3-hydroxypyridazine 97%

Supplier: Ambeed
Warning

A113501-100G A113501-10G A113501-25G A113501-500G A113501-5G
77108-124EA 50.47 USD
77108-124 77108-126 77108-128 77108-130 77565-990
6-Chloro-3-hydroxypyridazine 97%
6-Chloro-3-hydroxypyridazine

Precaución: For research use only.

Formula: C₄H₃ClN₂O
MDL Number: MFCD00233955
CAS Number: 19064-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR