Ethyl-p-toluate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-methylbenzoate, Ethyl-4-methylbenzoate, 4-Methylbenzoic acid ethyl ester, 4-Toluic acid ethyl ester, p-Toluic acid ethyl ester

A113321-100G A113321-10G A113321-25G A113321-500G A113321-5G
77108-114EA 37.14 USD
77108-114 77108-116 77108-118 77108-120 77108-122
Ethyl-p-toluate 97%
Ethyl-p-toluate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
MDL Number: MFCD00009117
CAS Number: 94-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR