3-Bromo-2-methox-6-methylypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3-Bromo-2-methoxy-6-methylpyridine

A112641-10G A112641-1G A112641-250MG A112641-25G A112641-5G
77108-088EA 237.65 USD
77108-088 77108-090 77108-092 77108-094 77108-096
3-Bromo-2-methox-6-methylypyridine 98%
3-Bromo-2-methox-6-methylypyridine

Precaución: for Research Use Only

Formula: C₇H₈BrNO
MW: 202.0485 g/mol
MDL Number: MFCD04112515
CAS Number: 717843-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR