(3-Chlorophenyl)(piperidin-4-yl)methanone 97%

Supplier: Ambeed
Warning

A112637-100MG A112637-1G A112637-250MG
77108-082EA 50.93 USD
77108-082 77108-084 77108-086
(3-Chlorophenyl)(piperidin-4-yl)methanone 97%
(3-Chlorophenyl)(piperidin-4-yl)methanone

Precaución: for Research Use Only

Formula: C₁₂H₁₄ClNO
MW: 223.7000 g/mol
MDL Number: MFCD04039472
CAS Number: 887354-02-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR