4-Bromo-7-fluoroquinoline 95%

Supplier: Ambeed
Warning

A112530-100MG A112530-1G A112530-250MG A112530-25G A112530-5G
77108-064EA 20.6 USD
77108-064 77108-066 77108-068 77108-070 77108-072
4-Bromo-7-fluoroquinoline 95%
4-Bromo-7-fluoroquinoline

Precaución: for Research Use Only

Formula: C₉H₅BrFN
MW: 226.045 g/mol
MDL Number: MFCD08063190
CAS Number: 1070879-29-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR