3-Chloroanisole 98%

Supplier: Ambeed
Warning

A112446-100G A112446-10G A112446-25G A112446-5G
77108-048EA 42.23 USD
77108-048 77108-050 77108-052 77108-054
3-Chloroanisole 98%
3-Chloroanisole

Precaución: for Research Use Only

Formula: C₇H₇ClO
MDL Number: MFCD00000591
CAS Number: 2845-89-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR