2-(3,5-Diphenyl-1H-pyrazol-1-yl)acetic acid 95%

Supplier: Ambeed
Warning

A112019-1G A112019-250MG
77108-032EA 176.11 USD
77108-032 77108-034
2-(3,5-Diphenyl-1H-pyrazol-1-yl)acetic acid 95%
2-(3,5-Diphenyl-1H-pyrazol-1-yl)acetic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₄N₂O₂
MW: 278.30 g/mol
MDL Number: MFCD00299897
CAS Number: 93323-67-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR