o-Tolylhydrazinium chloride 97%

Supplier: Ambeed

Synonyms: o-Tolylhydrazine hydrochloride, 1-(2-Methylphenyl)hydrazine hydrochloride, 2-Tolylhydrazine hydrochloride, 1-(2-Tolyl)hydrazine hydrochloride, 2-Methylphenylhydrazinium chloride, 2-Methylphenylhydrazine hydrochloride

A111609-100G A111609-10G A111609-1G A111609-25G A111609-5G
77107-984EA 55.62 USD
77107-984 77107-986 77107-988 77107-990 77107-992
o-Tolylhydrazinium chloride 97%
o-Tolylhydrazinium chloride

Precaución: for Research Use Only

Formula: C₇H₁₁ClN₂
MDL Number: MFCD00012925
CAS Number: 635-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR