2-Phthalimidopropionic acid 99%

Supplier: Ambeed
Danger

Synonyms: Phthalyl-DL-alanine, N-Phthaloyl-DL-alanine, 2-(1,3-Dioxoisoindolin-2-yl)propanoic acid

A111598-25G A111598-5G
77107-980EA 129.43 USD
77107-980 77107-982
2-Phthalimidopropionic acid 99%
2-Phthalimidopropionic acid

Precaución: for Research Use Only

Formula: C₁₁H₉NO₄
MW: 219.20 g/mol
MDL Number: MFCD00023061
CAS Number: 19506-87-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR