2,5-Bis(benzyloxy)benzoic acid 98%

Supplier: Ambeed
Warning

A111500-1G A111500-250MG A111500-5G
77107-974EA 57.29 USD
77107-974 77107-976 77107-978
2,5-Bis(benzyloxy)benzoic acid 98%
2,5-Bis(benzyloxy)benzoic acid

Precaución: for Research Use Only

Formula: C₂₁H₁₈O₄
MW: 334.37 g/mol
MDL Number: MFCD00156981
CAS Number: 67127-91-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR