1,3-Diphenylacetone 97%

Supplier: Ambeed
Warning

Synonyms: 1,3-Diphenylpropan-2-one, 1,3-Diphenyl-2-propanone

A111155-100G A111155-10G A111155-500G
77107-952EA 49.87 USD
77107-952 77107-954 77107-958
1,3-Diphenylacetone 97%
1,3-Diphenylacetone

Precaución: for Research Use Only

Formula: (C₆H₅)CH₂C(O)CH₂(C₆H₅)
MW: 210.28 g/mol
MDL Number: MFCD00004795
CAS Number: 102-04-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR