5-Fluoro-2-picoline hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-2-methylpyridine

A111061-1G A111061-25G A111061-5G
77107-938EA 24.72 USD
77107-938 77107-940 77107-942
5-Fluoro-2-picoline hydrochloride 98%
5-Fluoro-2-picoline hydrochloride

Precaución: for Research Use Only

Formula: C₆H₇ClFN
MW: 111.1169 g/mol
MDL Number: MFCD03095271
CAS Number: 31181-53-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR