Boc-D-2,4-diaminobutyric acid 95%

Supplier: Ambeed
Warning

Synonyms: Boc-D-Dab-OH

A110485-100MG A110485-250MG
77107-930EA 7.21 USD
77107-930 77107-932
Boc-D-2,4-diaminobutyric acid 95%
Boc-D-2,4-diaminobutyric acid

Precaución: for Research Use Only

Formula: C₉H₁₈N₂O₄
MDL Number: MFCD00798622
CAS Number: 80445-78-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR