5-Methylpyrazine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A110450-100G A110450-10G A110450-25G A110450-500G A110450-5G
77107-920EA 67.98 USD
77107-920 77107-922 77107-924 77107-926 77107-928
5-Methylpyrazine-2-carboxylic acid 98%
5-Methylpyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₆N₂O₂
MDL Number: MFCD00068241
CAS Number: 5521-55-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR