Anethole trithione 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(4-Methoxyphenyl)-3H-1,2-dithiole-3-thione, Felviten
A109759-10G
A109759-1G
A109759-25G
A109759-5G
77107-878EA
54.11
USD
77107-878
77107-880
77107-882
77107-884
Anethole trithione 98%
Anethole trithione
Precaución: for Research Use Only
Formula:
C₁₀H₈OS₃ MW: 240.3649 g/mol |
MDL Number:
MFCD00129751 CAS Number: 532-11-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...