(4-Fluoronaphthalen-1-yl)methanol 95%

Supplier: Ambeed
Warning

A109051-100MG A109051-1G A109051-250MG
77107-854EA 30.77 USD
77107-854 77107-856 77107-858
(4-Fluoronaphthalen-1-yl)methanol 95%
(4-Fluoronaphthalen-1-yl)methanol

Precaución: for Research Use Only

Formula: C₁₁H₉FO
MW: 176.18 g/mol
MDL Number: MFCD08059454
CAS Number: 79996-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR