Isothiazole-5-carboxylic acid 97%

Supplier: Ambeed
Warning

A108393-100MG A108393-1G A108393-5G A108393-250MG
77107-808EA 21.22 USD
77107-808 77107-810 77107-812 77592-162
Isothiazole-5-carboxylic acid 97%
Isothiazole-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₄H₃NO₂S
MW: 129.137 g/mol
MDL Number: MFCD07369964
CAS Number: 10271-85-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR