6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A108354-100G
A108354-1G
A108354-25G
A108354-5G
77107-800EA
71.07
USD
77107-800
77107-802
77107-804
77107-806
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 95%
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Precaución: for Research Use Only
Formula:
C₁₁H₁₆ClNO₂ |
MDL Number:
MFCD00012744 CAS Number: 2328-12-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...