(4-Butoxy-1H-pyrazolo[3,4-b]pyridin-5-yl)(2,6-difluoro-4-methylphenyl)methanone 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
BMS-265246
A107996-10MG
A107996-1MG
A107996-5MG
77107-794EA
55.17
USD
77107-794
77107-796
77107-798
(4-Butoxy-1H-pyrazolo[3,4-b]pyridin-5-yl)(2,6-difluoro-4-methylphenyl)methanone 98+%
(4-Butoxy-1H-pyrazolo[3,4-b]pyridin-5-yl)(2,6-difluoro-4-methylphenyl)methanone
Precaución: for Research Use Only
Formula:
C₁₈H₁₇F₂N₃O₂ MW: 345.34 g/mol |
MDL Number:
MFCD22420825 CAS Number: 582315-72-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...