Nitazoxanide 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(Acetyloxy)-N-(5-nitro-2-thiazolyl)benzamide, 2-((5-Nitrothiazol-2-yl)carbamoyl)phenyl acetate, NTZ
A107450-100G
A107450-100MG
A107450-10G
A107450-10MG
A107450-25G
A107450-500G
A107450-50MG
77107-754EA
64.89
USD
77107-754
77107-756
77107-758
77107-760
77107-762
77107-764
77107-766
Nitazoxanide 99%
Nitazoxanide
Precaución: for Research Use Only
Formula:
C₁₂H₉N₃O₅S |
MDL Number:
MFCD00416599 CAS Number: 55981-09-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...