2,5-Dichloro-4-pyridineboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dichloropyridine-4-boronic acid, 2,5-Dichloro-4-pyridylboronic acid

A107271-10G A107271-1G A107271-250MG A107271-5G
77107-738EA 108.22 USD
77107-738 77107-740 77107-742 77107-744
2,5-Dichloro-4-pyridineboronic acid 98%
2,5-Dichloro-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BCl₂NO₂
MW: 191.81 g/mol
MDL Number: MFCD06798254
CAS Number: 847664-64-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR