5-Chloro-2-fluorophenylacetonitrile 95%

Supplier: Ambeed
Warning

A107261-100G A107261-1G A107261-250MG A107261-25G A107261-5G
77107-728EA 242.04 USD
77107-728 77107-730 77107-732 77107-734 77107-736
5-Chloro-2-fluorophenylacetonitrile 95%
5-Chloro-2-fluorophenylacetonitrile

Precaución: for Research Use Only

Formula: C₈H₅ClFN
MDL Number: MFCD01631438
CAS Number: 75279-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR