2,3-Dimethoxybenzaldehyde 97%

Supplier: Ambeed
Warning

A107122-1000G A107122-100G A107122-10G A107122-25G A107122-500G A107122-5G
77107-716EA 143.17 USD
77107-716 77107-718 77107-720 77107-722 77107-724 77107-726
2,3-Dimethoxybenzaldehyde 97%
2,3-Dimethoxybenzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD00003309
CAS Number: 86-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR