1,4-Dibromo-2,5-bis(trifluoromethyl)benzene 95%

Supplier: Ambeed
Warning

Synonyms: 1,4-Bis(trifluoromethyl)-2,5-dibromobenzene, 1,4-Dibromo-2,5-bis(trifluoromethyl)benzene

A106983-1G A106983-5G
77107-714EA 43.5 USD
77107-714 77549-054
1,4-Dibromo-2,5-bis(trifluoromethyl)benzene 95%
1,4-Dibromo-2,5-bis(trifluoromethyl)benzene

Precaución: for Research Use Only

Formula: C₈H₂Br₂F₆
MW: 371.9 g/mol
MDL Number: MFCD15526888
CAS Number: 2375-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR