Disodium-2,7-naphthalenedisulfonate 98%

Supplier: Ambeed
Warning

Synonyms: 2,7-Naphthalenedisulphonic acid disodium salt, Sodium naphthalene-2,7-disulphonate, Ebert-Merz alpha-acid disodium salt

A106649-100G A106649-10G A106649-25G A106649-500G
77107-686EA 22.28 USD
77107-686 77107-688 77107-690 77107-692
Disodium-2,7-naphthalenedisulfonate 98%
Disodium-2,7-naphthalenedisulfonate

Precaución: for Research Use Only

Formula: C₁₀H₈O₆S₂
MW: 332.25 g/mol
MDL Number: MFCD00066365
CAS Number: 1655-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR