5-Chloro-6-methoxypyridine-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-6-methoxynicotinic acid

A106602-1G A106602-25G A106602-5G
77107-680EA 26.78 USD
77107-680 77107-682 77107-684
5-Chloro-6-methoxypyridine-3-carboxylic acid 98%
5-Chloro-6-methoxypyridine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₃
MDL Number: MFCD04972405
CAS Number: 884494-85-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR