4-(6-Bromopyridin-2-yl)benzoic acid 97%

Supplier: Ambeed
Warning

A106318-1G A106318-5G
77107-672EA 160.2 USD
77107-672 77107-674
4-(6-Bromopyridin-2-yl)benzoic acid 97%
4-(6-Bromopyridin-2-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₂H₈BrNO₂
MW: 278.1 g/mol
MDL Number: MFCD09753803
CAS Number: 928658-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR