CHIR-090 98+%

Supplier: Ambeed
Danger

Synonyms: N-((2S,3R)-3-Hydroxy-1-(hydroxyamino)-1-oxobutan-2-yl)-4-((4-(morpholinomethyl)phenyl)ethynyl)benzamide, N-[(2S,3R)-3-hydroxy-1-(hydroxyamino)-1-oxobutan-2-yl]-4-[2-[4-(morpholin-4-ylmethyl)phenyl]ethynyl]benzamide

A106028-10MG A106028-1MG A106028-2MG A106028-50MG A106028-5MG
77107-648EA 44.56 USD
77107-648 77107-650 77107-652 77107-654 77107-656
CHIR-090 98+%
CHIR-090

Precaución: for Research Use Only

Formula: C₂₄H₂₇N₃O5
MW: 437.5 g/mol
MDL Number: MFCD22665727
CAS Number: 728865-23-4
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR