2,4-Dichloro-5-fluoro-3-nitrobenzoic acid 97%

Supplier: Ambeed
Warning

A105665-10G A105665-25G
77107-566EA 6.37 USD
77107-566 77107-568
2,4-Dichloro-5-fluoro-3-nitrobenzoic acid 97%
2,4-Dichloro-5-fluoro-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₂Cl₂FNO₄
MW: 254 g/mol
MDL Number: MFCD03427137
CAS Number: 106809-14-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR