tert-Butyl 4-(cyanomethyl)-4-hydroxypiperidine-1-carboxylate 96%

Supplier: Ambeed
Warning

A105535-100MG A105535-250MG A105535-1G
77107-554EA 72.15 USD
77107-554 77107-556 77570-326
tert-Butyl 4-(cyanomethyl)-4-hydroxypiperidine-1-carboxylate 96%
tert-Butyl 4-(cyanomethyl)-4-hydroxypiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₀N₂O₃
MW: 240.3 g/mol
MDL Number: MFCD11111486
CAS Number: 774609-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR