N-Acetyl-L-methionine 95%

Supplier: Ambeed
Warning

Synonyms: N-Ac-Met-OH, N-Acetyl-L-Methionine, Ac-Met-OH NAM, Ac-Met-OH

A105058-100G A105058-500G A105058-10G A105058-25G A105058-1000G
77107-496EA 26.53 USD
77107-496 77107-498 77565-832 77570-300 77684-952
N-Acetyl-L-methionine 95%
N-Acetyl-L-methionine

Precaución: For research use only.

Formula: C₇H₁₃NO₃S
MW: 191.2 g/mol
MDL Number: MFCD00064441
CAS Number: 65-82-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR