4-Amino-2-chloro-5-nitrobenzotrifluoride 98%

Supplier: Ambeed
Warning

Synonyms: 5-Chloro-2-nitro-4-(trifluoromethyl)aniline

A104866-10G A104866-1G A104866-250MG A104866-25G A104866-5G
77107-478EA 118.45 USD
77107-478 77107-480 77107-482 77107-484 77107-486
4-Amino-2-chloro-5-nitrobenzotrifluoride 98%
4-Amino-2-chloro-5-nitrobenzotrifluoride

Precaución: for Research Use Only

Formula: C₇H₄ClF₃N₂O₂
MDL Number: MFCD01312277
CAS Number: 35375-74-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR