5-Iodo-2-methyl-1H-imidazole 97%

Supplier: Ambeed
Warning

A104828-10G A104828-1G A104828-250MG A104828-5G
77107-460EA 113.52 USD
77107-460 77107-462 77107-464 77107-466
5-Iodo-2-methyl-1H-imidazole 97%
5-Iodo-2-methyl-1H-imidazole

Precaución: for Research Use Only

Formula: C₄H₅IN₂
MW: 208 g/mol
MDL Number: MFCD15071119
CAS Number: 73746-45-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR