ML130 95%

Supplier: Ambeed
Warning

Synonyms: 1-Tosyl-1H-benzo[d]imidazol-2-amine, Nodinitib-1

A104782-10MG A104782-1MG A104782-25MG A104782-5MG
77107-452EA 89.12 USD
77107-452 77107-454 77107-456 77107-458
ML130 95%
ML130

Precaución: for Research Use Only

Formula: C₁₄H₁₃N₃O₂S
MW: 287.3369 g/mol
MDL Number: MFCD04322367
CAS Number: 799264-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR