alpha-Arbutin 99+%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxyphenyl alpha-D-Glucopyranoside, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(4-hydroxyphenoxy)tetrahydro-2H-pyran-3,4,5-triol

A104595-100G A104595-100MG A104595-10G A104595-1G A104595-25G A104595-500G A104595-5G
77107-438EA 212.18 USD
77107-438 77107-440 77107-442 77107-444 77107-446 77107-448 77107-450
alpha-Arbutin 99+%
alpha-Arbutin

Precaución: for Research Use Only

Formula: C₁₂H₁₆O₇
MDL Number: MFCD09838262
CAS Number: 84380-01-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR