1-(6-Methylpyridin-3-yl)-2-(4-(methylsulfonyl)phenyl)ethanone 97%

Supplier: Ambeed
Warning

A104580-100G A104580-10G A104580-1G A104580-250MG A104580-25G A104580-5G
77107-426EA 481.65 USD
77107-426 77107-428 77107-430 77107-432 77107-434 77107-436
1-(6-Methylpyridin-3-yl)-2-(4-(methylsulfonyl)phenyl)ethanone 97%
1-(6-Methylpyridin-3-yl)-2-(4-(methylsulfonyl)phenyl)ethanone

Precaución: for Research Use Only

Formula: C₁₅H₁₅NO₃S
MW: 289.34 g/mol
MDL Number: MFCD11110696
CAS Number: 221615-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR