(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone 97%

Supplier: Ambeed
Warning

A104453-10G A104453-1G A104453-25G A104453-5G A104453-100G
77107-408EA 85.94 USD
77107-408 77107-410 77107-412 77107-414 77570-272
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone 97%
(4R,5S)-(+)-4-Methyl-5-phenyl-2-oxazolidinone

Precaución: for Research Use Only

Formula: C₁₀H₁₁NO₂
MDL Number: MFCD00010845
CAS Number: 77943-39-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR