2-Bromo-5-fluoropyrimidine 95%

Supplier: Ambeed
Warning

A104410-100MG A104410-1G A104410-250MG A104410-5G
77107-400EA 10.31 USD
77107-400 77107-402 77107-404 77107-406
2-Bromo-5-fluoropyrimidine 95%
2-Bromo-5-fluoropyrimidine

Precaución: for Research Use Only

Formula: C₄H₂BrFN₂
MDL Number: MFCD09835164
CAS Number: 947533-45-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR