Ammonium benzenesulfonate 97%

Supplier: Ambeed
Warning

A103692-1G A103692-250MG A103692-25G A103692-5G
77107-364EA 8.24 USD
77107-364 77107-366 77107-368 77107-370
Ammonium benzenesulfonate 97%
Ammonium benzenesulfonate

Precaución: for Research Use Only

Formula: C₆H₉NO₃S
MW: 175.2100 g/mol
MDL Number: MFCD00191294
CAS Number: 19402-64-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR