3-Amino-2-methoxy-4-picoline 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methoxy-4-methylpyridin-3-amine

A102999-10G A102999-250MG A102999-5G
77107-306EA 311.91 USD
77107-306 77107-308 77684-938
3-Amino-2-methoxy-4-picoline 95%
3-Amino-2-methoxy-4-picoline

Precaución: for Research Use Only

Formula: C₇H₁₀N₂O
MW: 138.1671 g/mol
MDL Number: MFCD08277287
CAS Number: 76005-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR