3-Methylpyridin-2-ol 97%

Supplier: Ambeed
Warning

A102852-1G A102852-5G
77107-288EA 9.55 USD
77107-288 77107-290
3-Methylpyridin-2-ol 97%
3-Methylpyridin-2-ol

Precaución: for Research Use Only

Formula: C₆H₇NO
MW: 109.13 g/mol
MDL Number: MFCD00126917
CAS Number: 91914-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR