2,4,5-Trifluorobenzyl alcohol 98%

Supplier: Ambeed
Warning

A102841-10G A102841-1G A102841-25G
77107-284EA 30.91 USD
77107-284 77107-286 77684-937
2,4,5-Trifluorobenzyl alcohol 98%
2,4,5-Trifluorobenzyl alcohol

Precaución: for Research Use Only

Formula: C₇H₅F₃O
MDL Number: MFCD00061206
CAS Number: 144284-25-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR