Methyl 2-pivalamidoisonicotinate 95+%

Supplier: Ambeed
Warning

A102782-100MG A102782-1G A102782-250MG
77107-278EA 10.31 USD
77107-278 77107-280 77107-282
Methyl 2-pivalamidoisonicotinate 95+%
Methyl 2-pivalamidoisonicotinate

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂O₃
MDL Number: MFCD04039862
CAS Number: 1235666-16-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR