1-(tert-Butoxycarbonyl)-4-(4-methoxyphenyl)pyrrolidine-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A102216-100MG A102216-250MG
77107-234EA 72.15 USD
77107-234 77107-236
1-(tert-Butoxycarbonyl)-4-(4-methoxyphenyl)pyrrolidine-3-carboxylic acid 95%
1-(tert-Butoxycarbonyl)-4-(4-methoxyphenyl)pyrrolidine-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₅
MW: 321.37 g/mol
MDL Number: MFCD01631857
CAS Number: 851484-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR