Tetrabutylammonium fluoride hydrate 85%

Supplier: Ambeed
Danger

A100141-10G A100141-25G A100141-100G
77107-144EA 6.18 USD
77107-144 77107-146 77684-923
Tetrabutylammonium fluoride hydrate 85%
Tetrabutylammonium fluoride hydrate

Precaución: for Research Use Only

Formula: C₁₆H₃₆FN·H₂O
MDL Number: MFCD00149980
CAS Number: 22206-57-1
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR