N-Biotinyl-ethylenediamine trifluoroacetate salt ≥96.5% (by HPLC), Sigma-Aldrich®

Synonyms: N-(2-Aminoethyl)biotinamide trifluoroacetate salt

M-Clarity™ quality level = MQ100

08599-100MG 08599-500MG
220032-806EA 395.38 USD
220032-806 220032-807
N-Biotinyl-ethylenediamine trifluoroacetate salt ≥96.5% (by HPLC), Sigma-Aldrich®
N-Biotinyl-ethylenediamine trifluoroacetate salt

Nucleophilic biotinylation reagent for the conjugation of various substrates and their identification and quantification using biotin-binding fluorescent labels

Formula: C₁₂H₂₂N₄O₂S·C₂HF₃O₂
MW: 400.42 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11045268
CAS Number: 1217450-40-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR