2-(3-Fluoro-4-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%

Supplier: Ambeed
Warning

Synonyms: 3-Fluoro-4-(methylthio)phenylboronic acid pinacol ester

A991568-100MG A991568-1G A991568-250MG
77106-516EA 34 USD
77106-516 77106-518 77106-520
2-(3-Fluoro-4-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 98%
2-(3-Fluoro-4-(methylthio)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

Precaución: for Research Use Only

Formula: C₁₃H₁₈BFO₂S
MDL Number: MFCD22494876
CAS Number: 1351499-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR