4-[2-(2-Hydroxyethoxy)ethyl]morpholine 97%
Supplier: Ambeed
|
Warning
|
A990564-1G
A990564-250MG
A990564-5G
77106-510EA
53
USD
77106-510
77106-512
77106-514
4-[2-(2-Hydroxyethoxy)ethyl]morpholine 97%
4-[2-(2-Hydroxyethoxy)ethyl]morpholine
Precaución: for Research Use Only
Formula:
C₈H₁₇NO₃ |
MDL Number:
MFCD01095822 CAS Number: 3603-45-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...