2-Bromo-1-(5-chloro-2-thienyl)-1-ethanone 96%

Supplier: Ambeed
Danger

Synonyms: 2-Bromo-1-(5-chlorothiophen-2-yl)ethanone, 2-(Bromoacetyl)-5-chlorothiophene

A985383-250MG A985383-5G
77106-506EA 19.1 USD
77106-506 77106-508
2-Bromo-1-(5-chloro-2-thienyl)-1-ethanone 96%
2-Bromo-1-(5-chloro-2-thienyl)-1-ethanone

Precaución: for Research Use Only

Formula: C₆H₄BrClOS
MW: 239.52 g/mol
MDL Number: MFCD00829820
CAS Number: 57731-17-6
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR